Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSK1, p Ser380 Antibody (1024F) - Azide and BSA Free, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals MAB79672
Description
RSK1 Monoclonal specifically detects RSK1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RSK1 (p Ser380) | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 0.25 ug/ml, Immunocytochemistry 0.3 - 1 ug/ml | |
90kD, polypeptide 1), EC 2.7.11, EC 2.7.11.1,90 kDa ribosomal protein S6 kinase 1, HU-1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, p90RSK1, p90S6K, ribosomal protein S6 kinase alpha 1, ribosomal protein S6 kinase alpha-1, ribosomal protein S6 kinase, 90kD, polypeptide 1, ribosomal protein S6 kinase, 90kDa, polypeptide 1, Ribosomal S6 kinase 1, RSK, RSK-1, RSK1p90-RSK 1, S6K-alpha 1, S6K-alpha-1 | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
1024F | |
Unconjugated | |
PBS with No Preservative | |
RPS6KA1 | |
Phosphopeptide containing the human RSK1 S380 site Accession # Q15418 | |
100 μg | |
MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction | |
6195 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction