Learn More
RSK1 Rabbit anti-Human, Mouse, Unconjugated, Clone: ARC1096, Invitrogen™
Rabbit Monoclonal Antibody
Manufacturer:  Invitrogen MA535559
Catalog No. PIMA535559
Add to cart
Description
Description
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
RSK1
ARC1096
PBS with 50% glycerol and 0.2% sodium azide; pH 7.3
P18653, Q15418
90 kDa ribosomal protein S6 kinase 1; CLS; HU-1; HU-2; HU-3; ISPK-1; MAP kinase-activated protein kinase 1; MAP kinase-activated protein kinase 1a; MAPK-activated protein kinase 1; MAPK-activated protein kinase 1a; MAPKAP kinase 1c; Mapkapk1a; Mapkapk-1a; MAPKAPK1B; MAPKAPK1C; MAPKAPK-1c; MRX19; OTTHUMP00000004114; OTTMUSP00000010160; p90Rsk; p90-RSK; p90-RSK 1; p90-RSK 2; p90Rsk1; p90-Rsk1; p90RSK2; p90-RSK2; p90-RSK3; p90S6K; pp90RSK1; pp90RSK2; pp90RSK3; Ribosomal protein S6 kinase 2 alpha; Ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase II alpha; Ribosomal S6 kinase 1; RP11-393H10.3; RP11-492M19.2; RPS6KA; Rps6ka1; RSK; Rsk1; RSK-1; RSK2; RSK3; RSK-3; S6K-alpha; S6K-alpha-1; S6K-alpha2; S6K-alpha-2; S6K-alpha3; S6KII-alpha; wu:fc56h07; XELAEV_18012109mg; zgc:152654
Rabbit
IgG
100 μL
-20°C, Avoid Freeze/Thaw Cycles
Primary
20111, 6195
Western Blot
Unconjugated
RPS6KA1
Liquid
RPS6KA1
A synthesized peptide derived from human RSK1 p90
Protein A
RUO
Antibody
Monoclonal
Human, Mouse