Learn More
Showa Denko America Inc RSpak NN-814 DFS Item
Manufacturer:  Showa Denko America Inc F7008140
Polyhydroxymethacrylate packing material modified with sulfo groups supports multimode (reversed phase and cation exchange) analysis, ideal for the analysis of complex samples containing neutral and ionic substances, recommended for amino acids and carnitine
Catalog No. 50-161-3248