Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSPH3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$337.75 - $627.50
Specifications
Antigen | RSPH3 |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RSPH3 Polyclonal specifically detects RSPH3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RSPH3 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
83861 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:IIEVDMECQTDAFLDRPPTPLFIPAKTGKDVATQILEGELFDFDLEVKPVLEVLVGKTIEQSLLEVMEEEELANLRASQREYEELRN | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
radial spoke 3 homolog (Chlamydomonas), radial spoke head 3 homolog, radial spoke head protein 3 homolog, Radial spoke head-like protein 2, radial spoke protein 3, radial spokehead-like 2, RSHL2, RSP3dJ111C20.1 | |
RSPH3 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title