Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSPO1 Monoclonal Antibody (OTI11A9), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526680
Description
RSPO1 Monoclonal antibody specifically detects RSPO1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
RSPO1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q2MKA7 | |
RSPO1 | |
Human recombinant protein fragment corresponding to amino acids 21-263 of RSPO1 produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
OTI11A9 | |
Unconjugated | |
RSPO1 | |
CRISTIN3; cristin-3; cysteine-rich and single thrombospondin domain-containing protein 3; FLJ40906; hRspo1; mCristin-3; RGD1565558; Roof plate-specific spondin-1; RP11-566C13.1; RSPO; RSPO1; Rspondin; R-spondin; R-spondin 1; R-spondin homolog; R-spondin homolog (Xenopus laevis); R-spondin-1; thrombospondin type 1 domain containing gene Rspondin | |
Mouse | |
Affinity Chromatography | |
RUO | |
284654 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction