Learn More
RSPO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 177811AP
Catalog No. 50-173-1789
Add to cart
Description
Description
R-spondins signaling.
Specifications
Specifications
RSPO2
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6UXX9, Q8BFU0
CRISTIN2, hRspo2, R spondin 2, Roof plate specific spondin 2, RSPO2
Rabbit
IgG
150 μL
-20°C
Primary
239405, 340419, 500863
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RSPO2
Liquid
RSPO2
RSPO2 Fusion Protein Ag11216
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat