Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RSPO3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556307
Description
Immunogen sequence: PTNETRKCTV QRKKCQKGER GKKGRERKRK KPNKGESKEA IPDSKSLESS KEIPEQRENK QQQKKRKVQD KQKSVSVS Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 67%.
RSPO3 is a member of the R-spondin family and plays a role in the regulation of Wnt (wingless-type MMTV integration site family)/beta-catenin and Wnt/planar cell polarity (PCP) signaling pathways, which are involved in development, cell growth and disease pathogenesis. RSPO3 has been shown to be involved in regulating the balance between angioblast and blood cell specification during embryonic vasculogenesis and angiogenesis and may be involved in tumor development.
Specifications
RSPO3 | |
Polyclonal | |
Unconjugated | |
RSPO3 | |
2810459H04Rik; AW742308; Cabriolet; cristin 1; Cristin1; cristin-1; cysteine-rich and single thrombospondin domain-containing protein 1; hPWTSR; hRspo3; Nucleopondin; protein with TSP type-1 repeat; PWTSR; RGD1563246; Roof plate-specific spondin-3; Rspo3; R-spondin 3; R-spondin 3 homolog; R-spondin 3 homolog (Xenopus laevis); R-spondin 3-like protein; R-spondin-3; Thrombospondin type-1 domain-containing protein 2; thrombospondin, type I, domain 2; thrombospondin, type I, domain containing 2; Thsd2; Unknown (protein for MGC:137973) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84870 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BXY4 | |
RSPO3 | |
Recombinant protein corresponding to Human RSPO3. Recombinant protein control fragment (Product #RP-94892). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction