Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSV Fusion Protein Human anti-Virus, DyLight 594, Clone: RSHZ19 (Felvizumab), Novus Biologicals™
Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275190DL594
Description
RSV Fusion Protein Monoclonal specifically detects RSV Fusion Protein in Viral samples. It is validated for ELISA.Specifications
RSV Fusion Protein | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
This antibody was prepared by the humanization of an F protein-specific murine MAb, RSV19. More specifically, molecular techniques were used to insert the complementarity determining regions from murine RSV19 into a human IgG1 heavy and k light chain-variable domain framework. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA | |
RSHZ19 (Felvizumab) | |
ELISA | |
Human | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title