RSVA 405, Tocris™

Manufacturer: R&D Systems 5138/50

Quantity 50mg

 View more versions of this product

Catalog No. 51-385-0

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 140405-36-3
    Molecular Formula C17H20N4O2
    Formula Weight 312.37
    Synonym 2-[[4-(Diethylamino)-2-hydroxyphenyl]methylene]hydrazide-4-pyridinecarboxylic acid
    Chemical Name or Material RSVA 405

    Specifications

    Formula Weight 312.37
    Quantity 50mg
    Synonym 2-[[4-(Diethylamino)-2-hydroxyphenyl]methylene]hydrazide-4-pyridinecarboxylic acid
    Molecular Formula C17H20N4O2
    CAS 140405-36-3
    Recommended Storage Store at -20°C
    Chemical Name or Material RSVA 405

    Store at -20°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit