Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSVA 405, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
AMP-activated protein kinase (AMPK) activator (EC50 = 1 μM); activates AMPK indirectly through a CaMKKβ-dependent mechanism. Also inhibits LPS-induced STAT3 phosphorylation (IC50 = 0.5 μM). Induces autophagy and promotes Aβ degradation in vitro. Also inhibits adipogenesis in vitro and reduces weight gain in diet-induced obese mice.
Specifications
Quantity | 10 mg |
Formula Weight | 312.37 |
Chemical Name or Material | RSVA 405 |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction