Learn More
Rat IgG2b kappa, Biotin, clone: eB149/10H5, Isotype Control, eBioscience
Rat Isotype Control
Manufacturer: Invitrogen 13403182
Description
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin.Applications Reported: Biotin Rat IgG2b Isotype Control has been reported for use in immunohistochemistry, immunocytochemistry and flow cytometric analysis.Applications Tested: Biotin Rat IgG2b Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. Use at the same concentration as the experimental antibody.Filtration: 0.2µm post-manufacturing filtered.Specifications
Rat IgG2b kappa | |
Control | |
0.5mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
IgG2, IgG2b, IgG2bk | |
IgG2b, kappa | |
100μg | |
Store at 2-8°C. Do not freeze. | |
Monoclonal |
Control, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
eB149/10H5 | |
Biotin | |
Liquid | |
Rat | |
Affinity chromatography | |
RUO | |
Isotype Control | |
Not Applicable |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.