Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RtcB Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 198091AP

Catalog No. 50-173-2551


Add to cart

Description

Description

This antibody specifically recognizes endogenous RTCB protein.

The function of this protein remains unknown.
Specifications

Specifications

RtcB
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99LF4, Q9Y3I0
C22orf28, FAAP, HSPC117, RtcB
Rabbit
IgG
150 μL
-20°C
Primary
28088, 51493
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
RTCB
Liquid
Rtcb
RtcB-Specific Fusion Protein Ag13836
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit