Learn More
Novus Biologicals Rat Collagen X ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP275828
Description
The Rat Collagen X ELISA Kit (Colorimetric) measures rat Collagen X in serum, plasma and other biological fluids.Specifications
0.31 - 20 ng/mL | |
ELISA | |
0.19 ng/mL | |
Collagen X | |
1 Kit |
This kit recognizes Rat COL10 in samples. No significant cross-reactivity or interference between Rat COL10 and analogues was observed. | |
ELISA Kit (Colorimetric) | |
Storage is content dependent. | |
Rat | |
Biological Fluids, Plasma, Serum |