Learn More
Aviva Systems Biology Corp RSPH14 Antibody - middle region (ARP55056_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55056_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RTDR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RTDR1. Alternate names: MGC16968ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human
Catalog No. 50-014-61410