Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTDR1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548540

Catalog No. PA548540

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a protein with no known function but with slight similarity to a yeast vacuolar protein. The gene is located in a region deleted in pediatric rhabdoid tumors of the brain, kidney and soft tissues, but mutations in this gene have not been associated with the disease. [provided by RefSeq, Jul 2008]
Specifications

Specifications

RTDR1
Polyclonal
PBS with 0.09% sodium azide
Q9UHP6
Rhabdoid tumor deletion region protein 1, RTDR1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27156
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RSPH14
Liquid
RSPH14
KLH conjugated synthetic peptide between 17-45 amino acids from the N-terminal region of human RTDR1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit