Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTEL1 Rabbit anti-Human, Mouse, Alexa Fluor™ 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP222360AF350

Catalog No. NP222360A30


Add to cart

Description

Description

RTEL1 Polyclonal antibody specifically detects RTEL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

RTEL1
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
C20orf41, regulator of telomere elongation helicase 1, regulator of telomere length
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Affinity Purified, Conjugated
RTEL1
A genomic peptide made to an internal region of the human RTEL1 protein (within residues 800-1000). [Swiss-Prot Q9NZ71]
Immunogen affinity purified
Cancer, DNA Repair
Primary
51750.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit