Learn More
RTEL1 Rabbit anti-Human, Mouse, Alexa Fluor™ 594, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222360AF594
Catalog No. NP222360A94
Add to cart
Description
Description
RTEL1 Polyclonal antibody specifically detects RTEL1 in Human,Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
RTEL1
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
RTEL1
A genomic peptide made to an internal region of the human RTEL1 protein (within residues 800-1000). [Swiss-Prot Q9NZ71]
0.1 mL
Primary
51750
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
C20orf41, regulator of telomere elongation helicase 1, regulator of telomere length
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
Human, Mouse