Learn More
RTF1 Rabbit anti-Drosophila, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA540081
Catalog No. PA540081
Add to cart
Description
Description
PAF is a five-subunit protein complex composed of Paf1, Cdc73, Leo1, Rtf1 and Ctr9, which was first purified from yeast. The yeast PAF (yPAF) complex interacts with RNA polymerase II and coordinates the setting of histone marks associated with active transcription. The human PAF (hPAF) complex shares four subunits with yPAF (hCtr9, hPaf1, hLeo1 and Cdc73), but contains an additional higher eukaryotic-specific subunit, hSki8. In addition to coordinating events during transcription (initiation, promoter clearance and elongation); PAF also coordinates events in RNA quality control.
Specifications
Specifications
RTF1
Polyclonal
Unconjugated
Rtf1
Liquid
RTF1
Recombit drosophila Rtf1 (Rtf1, Paf1/RNA polymerase II complex component, homolog) protein.
Protein G
RUO
Antibody
Polyclonal
Drosophila
ChIP assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
1.8 mg/mL
PBS with 0.05% Proclin 300, 0.05% sodium azide; pH 7.4
Q9W261
RNA polymerase-associated protein RTF1 homolog, KIAA0252
Rabbit
IgG
50 μg
-20°C or -80°C if preferred
Primary
37554