Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTF1, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA540066

Catalog No. PA540066

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PAF is a five-subunit protein complex composed of Paf1, Cdc73, Leo1, Rtf1 and Ctr9, which was first purified from yeast. The yeast PAF (yPAF) complex interacts with RNA polymerase II and coordinates the setting of histone marks associated with active transcription. The human PAF (hPAF) complex shares four subunits with yPAF (hCtr9, hPaf1, hLeo1 and Cdc73), but contains an additional higher eukaryotic-specific subunit, hSki8. In addition to coordinating events during transcription (initiation, promoter clearance and elongation); PAF also coordinates events in RNA quality control.

Specifications

Specifications

RTF1
Polyclonal
whole serum with 0.05% sodium azide
Q9W261, Q92541
RNA polymerase-associated protein RTF1 homolog, KIAA0252
Rabbit
IgG
Store at -20°C. Avoid freeze/thaw cycles.
Primary
23168
Chromatin Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
RTF1
Serum
RTF1
Recombinant human drosophila Rtf1 (Rtf1, Paf1/RNA polymerase II complex component, homolog) protein.
100μL
Antibody
Polyclonal
Drosophila, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit