Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RTKN Antibody (OTI2E9), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273950AF488
Description
RTKN Monoclonal specifically detects RTKN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RTKN | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RTKN | |
Full length recombinant protein of human RTKN (NP_149035) produced in HEK293T cells. | |
0.1 mL | |
6242.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2E9 | |
Western Blot, Immunohistochemistry | |
B5, rhotekin, RTKN1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction