Learn More
RTN1 (Isoform RTN1-C) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150481AP
Catalog No. 50-172-9931
Add to cart
Description
Description
This antibody is raised against the full-length protein of human RTN1-C.
Reticulon-1 was formerly designated NSP for Neuroendocrine-Specific-Protein, because it is specifically expressed in neural and neuroendocrine tissues. The NSP-gene has been mapped by fluorescence in situ hybridization to human chromosome 14q21-q22. The NSP-gene encodes three overlapping proteins, i. e. reticulon-1A (NSP-A), reticulon-1B (NSP-B), and reticulon-1C (NSP-C). These proteins were found to be anchored to membranes of the endoplasmic reticulum through their common carboxy-terminal regions. Reticulon-1A is a protein with a molecular weight of ~135 kDa, which occurs in various isoforms presumably depending on the degree of phosphorylation of serine residues. In lung cancer diagnosis Reticulon-1A appeared to be a reliable marker for the detection of neuroendocrine differentiation, since most of the small cell lung carcinoma (SCLC) and carcinoid tumors showed expression of Reticulon-1A. Reticulon-1B is a phosphoprotein with a MW of 45 kDa and is restricted to the lung cancer cell line NCI-H82. Reticulon-1B is so far not found in human tissues. Reticulon-1C is a protein with a MW of 23 kDa which is not phosphorylated and is found with Reticulon-1A in SCLC (cell lines) and not in non-SCLC (cell cultures).
Specifications
Specifications
RTN1 (Isoform RTN1-C)
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16799, Q64548, Q8K0T0
NSP, NSP-C, reticulon 1, RTN1, RTN1-C
Rabbit
IgG
150 μL
-20°C
Primary
104001, 116644, 6252
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RTN1
Liquid
RTN1
RTN1 (Isoform RTN1-C) Fusion Protein Ag7031
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat