Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTN1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP258934

Catalog No. NBP258934


Add to cart

Description

Description

RTN1-A/NSP Polyclonal antibody specifically detects RTN1-A/NSP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

RTN1
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Neuroendocrine-specific proteinNSPMGC133250, reticulon 1, reticulon-1
Rabbit
IgG
100 ul
Neuroscience
Primary
6252
Human
Immunocytochemistry, Immunofluorescence
Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Affinity Purified
RTN1
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AYKYIDITRPEEVKHQEQHHPELEDKDLDFKNKDTDISIKPEGVREPDKPAPVEGKIIKDHLLEESTFAPYIDDLSEEQRRAPQITTPV
Affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only