Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RTN4IP1 Mouse anti-Human, Clone: OTI1B2, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA502543
Description
This gene encodes a novel mitochondrial protein that interacts with reticulon 4, which is a potent inhibitor of regeneration following spinal cord injury. The interaction of reticulon 4 with mitochondrial proteins may provide insight into the mechanisms for reticulon-induced inhibition of neurite growth.Specifications
RTN4IP1 | |
Monoclonal | |
0.56 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8WWV3 | |
NIMP, OPA10 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
84816 |
Flow Cytometry, Western Blot | |
OTI1B2 | |
Unconjugated | |
RTN4IP1 | |
Liquid | |
RTN4IP1 | |
Full length human recombit protein of human RTN4IP1 produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title