Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTN4IP1 Mouse anti-Human, Clone: OTI3B2, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502544

Catalog No. 50-167-3594


Add to cart

Description

Description

This gene encodes a novel mitochondrial protein that interacts with reticulon 4, which is a potent inhibitor of regeneration following spinal cord injury. The interaction of reticulon 4 with mitochondrial proteins may provide insight into the mechanisms for reticulon-induced inhibition of neurite growth.
Specifications

Specifications

RTN4IP1
Monoclonal
0.38 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8WWV3
NIMP, OPA10
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84816
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI3B2
Unconjugated
RTN4IP1
Liquid
RTN4IP1
Full length human recombit protein of human RTN4IP1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit