Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rtn4r, anti-Rat, Clone: M5, Abnova™

Monoclonal Antibody

Manufacturer:  Abnova Corporation RAB00093

Catalog No. 89-032-436


Add to cart

Specifications

Specifications

Rtn4r
M5
ELISA (0.1 ug/mL) Flow Cytometry (1 ug/mL) Immunoprecipitation (10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
reticulon 4 receptor
NgR|NgR1|Nogor
Original antibody is raised against His6 tagged fragment corresponding to amino acids 278-473 of rat Rtn4r.
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
113912
Human, Rat
ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Liquid
Rtn4r
IgG, kappa
RUO
Antibody
Monoclonal
This antibody recognizes human and rat NgR1 with a Kd of 2.9 nM for both species (bivalent binding).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit