Learn More
RTP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 189731AP
Catalog No. 50-173-2297
Add to cart
Description
Description
Specifically promotes functional cell surface expression of olfactory receptors, but not of other GPCRs.
Specifications
Specifications
RTP1
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P59025, Q8C8C1
RTP1
Rabbit
IgG
150 μL
-20°C
Primary
132112, 239766, 288006
Western Blot
Unconjugated
RTP1
Liquid
Rtp1
RTP1 Fusion Protein Ag6934
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat