Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ RTP4 Human Recombinant Protein

Highly purified. Generating reliable and reproducible results.

$436.00 - $1298.00

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-Page
Formulation Phosphate buffered saline (pH7.4) containing, 20% glycerol, 1mM DTT
Gene ID (Entrez) 64108
Molecular Weight TMW: 27.8kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25191501
View Documents
Novus Biologicals™
NBP2519150.1MG
0.1mg
Each for $436.00
Add to cart
 
NBP25191505
View Documents
Novus Biologicals™
NBP2519150.5MG
0.5mg
Each for $1,298.00
Add to cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-224 of Human RTP4 The Recombinant Human RTP4 Protein is derived from E. coli. The Recombinant Human RTP4 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.25mg/mL
Phosphate buffered saline (pH7.4) containing, 20% glycerol, 1mM DTT
TMW: 27.8kDa
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
RTP4
Yes
SDS-Page
64108
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
IFRG28receptor transporting protein 4, receptor (chemosensory) transporter protein 4,28kD interferon responsive protein, receptor transporter protein 4, receptor-transporting protein 4,28 kDa interferon-responsive protein
Unlabeled
SDS
Product Certifications

For research use only and is not approved for use in humans or in clinical diagnosis

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit