Learn More
RTRAF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA592009
Description
Immunogen sequence: MFRRKLTALD YHNPAGFNCK DETEFRNFIV WLEDQKIRHY KIEDRGNLRN IHSSDWPKFF EKYLRDVNCP FKIQDRQEAI DWLLGLAVRL EYGDNAEKYK DLVPDNSKTA DNATKNAEPL INLDVNNPDF KAGVMALANL LQIQRHDDYL VMLKAIRILV QERLTQDAVA KANQTKEGLP VALDKHILGF DTGDAVLNEA AQILRLLHIE ELRELQTKIN EAIVAVQAII ADPKTDHRLG KVGR; Positive Samples: Jurkat, 293T; Cellular Location: Cytoplasm, Nucleus, centrosome, cytoskeleton, cytosol, microtubule organizing center, perinuclear region
RNA-binding protein involved in modulation of mRNA transcription by Polymerase II. Component of the tRNA-splicing ligase complex and is required for tRNA ligation. May be required for RNA transport.; (Microbial infection) In case of infection by influenza virus A (IVA), is involved in viral replication.Specifications
C14orf166 | |
Polyclonal | |
Unconjugated | |
RTRAF | |
RTRAF | |
Recombit fusion protein containing a sequence corresponding to amino acids 1-244 of human C14orf166 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
2.79 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y224 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51637 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.