Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Rtx™-5 Capillary Columns, 0.25 mm I.D.

General-Purpose Columns for Drugs, Solvent Impurities, Pesticides, Hydrocarbons, PCB Congeners, Essential Oils, Semivolatiles
Specifications
Diameter (Metric) Inner | 0.25 mm |
---|---|
For Use With (Application) | Drugs, Solvent Impurities, Pesticides, Hydrocarbons, PCB Congeners (e.g., Aroclor Mixes), Essential Oils, Semivolatiles |
Stationary Phase | Crossbond Diphenyl Dimethyl Polysiloxane |
USP Type | G27, G36 |
Max. Temperature | 350°C |
Catalog Number | Mfr. No. | Film Thickness | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Film Thickness | Length (Metric) | Price | Quantity & Availability | ||||
06-610-646
![]() |
Restek™
10205 |
0.1 μm | 15 m |
Each for $398.00
|
N/A |
06-610-650
![]() |
Restek™
10211 |
0.1 μm | 60 m |
Each for $1,129.00
|
N/A |
06-610-654
![]() |
Restek™
10223600 |
0.25 μm | 30 m |
Pack of 6 for $3,210.00
|
N/A |
06-610-656
![]() |
Restek™
10226600 |
0.25 μm | 60 m |
Pack of 6 for $5,595.00
|
N/A |
06-610-658
![]() |
Restek™
10235 |
0.5 μm | 15 m |
Each for $380.00
|
N/A |
Description
- Removes all residual catalysts and low molecular weight fragments from the Rtx™-5 polymer
- Provides a tight, unimodal distribution
- Extremely low bleed
- For Drugs, Solvent Impurities, Pesticides, Hydrocarbons, PCB Congeners (e.g., Aroclor Mixes), Essential Oils, Semivolatiles
Specifications
0.25 mm | |
Crossbond Diphenyl Dimethyl Polysiloxane | |
350°C | |
Low |
Drugs, Solvent Impurities, Pesticides, Hydrocarbons, PCB Congeners (e.g., Aroclor Mixes), Essential Oils, Semivolatiles | |
G27, G36 | |
Rtx™-5 | |
Fused Silica |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title