Learn More
Restek Rtx-DHA Columns
Excellent responses and peak symmetry for polar oxygenates
$1118.00 - $1911.00
Specifications
For Use With (Application) DHA methods
Material Dimethyl polysiloxane
Type Capillary column
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
06-610-641
View Documents
Restek
10147
0.2 mm 0.5 μm 50 m
Each for $1,118.00
Add to cart
06-610-642
View Documents
Restek
10148
0.25 mm 0.5 μm 100 m
Each for $1,343.00
Add to cart
06-610-643
View Documents
Restek
10149
0.25 mm 1 μm 150 m
Each for $1,911.00
Add to cart
Description
Description
  • Columns meet or exceed all ASTM D6730-01 and CAN/CGSB 3.0 No. 14.3-99 method guidelines
    • Test report for method D6730 supplied with each column
  • Stable to 340 °C
Specifications
Specifications
DHA methods
Dimethyl polysiloxane
Capillary column