Learn More
Argos Technologies Manual Pipettes
Pinch and release valve system allows for single-hand operation.
Description
- For aspiration and delivery of lab liquids
- Pipetting bulbs manufactured from natural rubber
- For use with serological pipettes up to 50mL
- Each valve has chemically resistant stainless-steel ball bearing
- For use with
Specifications
serological pipettes up to 50mL | |
Manual pipette | |
Red | |
Natural rubber |