Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubbermaid™ 1/2 Glutton™ Recycling Station

High-capacity, all-in-one centralized solution for efficient waste sortation

Manufacturer:  Rubbermaid™ 1792340

 View more versions of this product

Catalog No. 18-367-011


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

35.4 in.
Plastic
46 gal., 174.1L
26.8 x 23.6 x 35.4 in. (68.1 x 59.9 x 89.9cm)
One Glutton receptacle, two Slim Jim™ receptacles, one lid frame, choice of three opening shapes: circle, slot, and square, label pack, 2 openings
68.1cm
23.6 in.
46 gal.
89.9cm
Recycling Station
Dark Green
For efficient waste sortation
26.8 in.
Rectangular
59.9cm
174.1L
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit