Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubbermaid™ BRUTE™ Round Container

Rubbermaind BRUTE™ Round Container features all-plastic seamless construction for added durability.

Manufacturer:  Rubbermaid™ FG264300GRAY

 View more versions of this product

Catalog No. 19-076-521

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Easy to clean; withstand bumps and kicks, and won't rust, chip, or peel
  • Reinforced rims add strength and durability
  • Built-in handles aid lifting and allow easy, no-jam nesting
  • Double-ribbed base ensures stability and increased dragging capacity
  • Snap-on lids contain odors and allow secure, stable stacking (sold separately, call your Fisher Safety Customer Service Representative for ordering information)

Certifications

Containers are USDA Meat and Poultry Equipment Group Listed.

Compliance

Assist in complying with HACCP guidelines.

Specifications

Specifications

166.55 L
Snap-on
60.96 cm
80.01 cm
Round
Gray
Safety
USDA, HACCP
24 in.
31.5 in.
Built in handles, reinforced rim
44 gal.
LLDPE
Container
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit