Rubbermaid™ HYGEN™ MF Flexi Frame Mops

Premium split nylon/polyester blend microfiber provides optimal damp mopping performance

Manufacturer: Rubbermaid™ FGQ87100BL00

 View more versions of this product

Catalog No. 19-167-282

  Check Availability
Add to Cart

    Specifications

    Height (English) 0.86 in.
    Length (English) 8.85 in.
    Width (English) 5.86 in.
    Color Blue
    Material MicroFiber
    Product Type Glass Mop Cover
    Model Glass mop

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit