Learn More
Rubbermaid™ HYGEN™ MF Flexi Frame Mops
Premium split nylon/polyester blend microfiber provides optimal damp mopping performance
Manufacturer:  Rubbermaid™ FGQ85600GR00
 View more versions of this product
Catalog No. 19-167-280
Add to cart
Description
Description
  • Sustainable attributes: Green cleaning and LEED cleaning
  • Up to 500 launderings when washed without bleach
  • Up to 200 launderings when washed with bleach
  • Specially designed microfiber for cleaning glass
  • HYGEN superior quality microfiber
  • Bleach safe
Specifications
Specifications
Dust mop
MicroFiber
0.25 in.
5.87 in.
Green
Dust Mop Cover
8.87 in.