Learn More
Rubbermaid™ Round BRUTE™ Containers
All-plastic seamless construction
Manufacturer:  Rubbermaid™ RCP263200GY
 View more versions of this product
Catalog No. 19-046-669
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Easy to clean; withstands bumps and kicks; will not rust, chip or peel
  • Reinforced rims add strength and durability
  • Built-in handles aid lifting and allow easy, no-jam nesting
  • Double-ribbed base ensures stability and increased dragging capacity
  • Snap-on lids (sold separately) contain odors and allow secure, stable stacking
Order Info
Universal dolly for all sizes of containers available separately (19-046-694).
Certifications
Gray, white, and yellow containers are USDA Meat and Poultry Equipment Group Listed
Compliance
Assist in complying with HACCP guidelines.
Specifications
Specifications
121 L
Snap-on
55.88 cm
56.51 cm
32 gal.
Polyethylene
Double Ribbed Base Waste Container
NSF, USDA, HACCP
22 in.
22.25 in.
Reinforced rim
Gray
Safety
Documents
Accessories