Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubbermaid™ Slim Jim™ Recycling Containers

Convenient central collection site for multiple workstations

Manufacturer:  Rubbermaid™ FG354075BLUE

 View more versions of this product

Catalog No. 19-813-107

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications Description & Specifications

Specifications

23 gal., 87.1L
20 x 11 x 30 in. (50.8 x 27.9 x 76.2cm)
For multiple workstations
20 in.
Rectangular
27.9cm
87.1L
76.2cm
Recycling Container
Blue, White
For use in areas of high paper generation, such as near copiers, printers, and in mailrooms
No venting channels
50.8cm
11 in.
23 gal.
30 in.
Plastic
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit