Learn More
Rubbermaid™ Tandem BRUTE™ Dolly
Manufacturer:  Rubbermaid™ FG264600BLA
Catalog No. 19-156-169
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Doubles productivity when collecting waste; Opens flat to accommodate two Brute containers at once; Folds up completely for easy storage.
  • Designed to fit a combination of different size Brute containers. Doubles productivity in collection of recyclables and general waste. Innovative center-hinge design allows dolly to be laid flat to accommodate two containers simultaneously, folded at a 90° angle for a single container, or folded up completely for easy storage. Heavy-duty 8 in. dia. (20.3cm) wheels and swivel casters offer exceptional mobility for maneuvering through the workplace.
Order Info
Tandem dolly measures 45L × 20 1/4W × 8 in.H (114 × 51 × 20.3cm). Capacity: 200 lb. (91kg) per dolly, or 400 lb. (181kg) total. Black.