Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubbermaid™ Trades Cart / Service Bench

Move productivity right to the worksite with the Rubbermaid Trade Cart/Service Bench. Its flat top surface, placed at an ergonomic height, provides a convenient work area.

Manufacturer:  Rubbermaid™ FG618000BLA

Catalog No. 11-926-125


Add to cart

Description

Description

  • Four compartmentalized boxes store assorted parts and hardware, and two storage bins hold essential hand tools and supplies
  • Integrated handle for maneuverability
  • Moves easily over hard surfaces and through elevators and doorways on 5 in. dia. (12.7cm) casters
  • Available with open bottom shelf or with fully enclosed lockable storage compartment
  • Load capacities: Top shelf: 180 lb. (81.6kg), bottom shelf: 150 lb. (68kg)
  • Dimensions: 19 1/4L x 38W x 33 3/8 in.H (49 x 97 x 85cm)
Specifications

Specifications

2 parts boxes and 4 parts bins
Trades cart
PP (Polypropylene)
68 to 81.6kg
84.8cm
96.5cm
38 x 19.25 x 33.37 in. (96.5 x 48.9 x 84.8cm)
149.7kg
33.37 in.
19.25 in.
150 to 180 lb.
Black
38 in.
48.9cm
330 lb.
4 Compartmentalized boxes, 2 parts boxes and 4 parts bins, integrated handle, open bottom shelf or with fully enclosed lockable storage compartment
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit