Alfa Aesar™ Rubidium bromide, 99.8% (metals basis)

Manufacturer: Alfa Aesar™ 1289114

Quantity 25g

 View more versions of this product

Catalog No. AA1289114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-39-1
    Formula Weight 165.38
    MDL Number MFCD00011186
    Merck Index 14,8283
    Chemical Name or Material Rubidium bromide

    Specifications

    Formula Weight 165.38
    Odor Odorless
    Sensitivity Hygroscopic
    MDL Number MFCD00011186
    CAS 7789-39-1
    Merck Index 14,8283
    Boiling Point 1340°C
    Melting Point 693°C
    Physical Form Crystalline
    Refractive Index 1.5530
    Solubility Information Soluble in water
    TSCA TSCA
    Quantity 25g
    Chemical Name or Material Rubidium bromide
    Assay 99.8%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit