Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Rubidium fluoride, 99.1% (metals basis)

Manufacturer:  Alfa Aesar™ 4714409

Quantity 10g

 View more versions of this product

Catalog No. AA4714409


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13446-74-7
104.47
UN3288
RbF
MFCD00011191
Rubidium fluoride

Specifications

13446-74-7
RbF
MFCD00011191
3.557g/mL
Hygroscopic
795°C
Rubidium fluoride
Crystalline
104.47
UN3288
1.398
White
10g
99.1%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : VL8740000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit