Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Rubidium fluoride hydrate, 99%

Click to view available options
Quantity:
10g
50g
16422-67-6
FH2ORb
122.481
MFCD00149834
CTSZMVONLYPOKW-UHFFFAOYSA-M
rubidium fluoride hydrate,f.rb.h2o,acmc-1c7r7,rubidium fluoride,hydrate 8ci,rubidium fluoride-water 1/1/1,rubidium 1+ ion hydrate fluoride
57370370
rubidium(1+);fluoride;hydrate
O.[F-].[Rb+]
16422-67-6
FH2ORb
122.481
MFCD00149834
CTSZMVONLYPOKW-UHFFFAOYSA-M
rubidium fluoride hydrate,f.rb.h2o,acmc-1c7r7,rubidium fluoride,hydrate 8ci,rubidium fluoride-water 1/1/1,rubidium 1+ ion hydrate fluoride
57370370
rubidium(1+);fluoride;hydrate
O.[F-].[Rb+]
Applications
Raw material used in the perparation of rubidium metal.

Solubility
Soluble in water.

Notes
Store in a cool, dry, conditions in well sealed container.
UN Number UN3288
Solubility Information Soluble in water.
Formula Weight 104.47 (Anhydrous)
Percent Purity 99%
Odor Odorless
Quantity 50g
Chemical Name or Material Rubidium fluoride hydrate

GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c

H301+H311-H315-H319-H330-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 236-603-2

RTECSNumber : VL8740000

TSCA : Yes

Recommended Storage : Ambient temperatures