Alfa Aesar™ Rubidium fluoride, Puratronic™, 99.975% (metals basis)

Manufacturer: Alfa Aesar™ 1287304

Quantity 2g

 View more versions of this product

Catalog No. AA1287304

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13446-74-7
    Linear Formula RbF
    Formula Weight 104.47
    MDL Number MFCD00011191
    UN Number UN3288
    Chemical Name or Material Rubidium fluoride
    Grade Puratronic™

    Specifications

    Formula Weight 104.47
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Sensitivity Hygroscopic
    UN Number UN3288
    Color White
    Melting Point 795°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Odor Odorless
    Physical Form Crystalline
    Refractive Index 1.398
    CAS 13446-74-7
    Health Hazard 1 Danger
    Density 3.557g/mL
    TSCA TSCA
    MDL Number MFCD00011191
    Quantity 2g
    RTECS Number VL8740000
    Linear Formula RbF
    Chemical Name or Material Rubidium fluoride
    Assay 99.975%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H301-H311-H331-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit