Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Rubidium hexafluoroarsenate(V), 98% (metals basis)
Click to view available options
Quantity:
5g
25g
Specifications
UN Number | UN1557 |
Solubility Information | Soluble in water. |
Formula Weight | 359.92 |
Physical Form | Powder |
Percent Purity | 98% |
Quantity | 5g |
Chemical Name or Material | Rubidium hexafluoroarsenate(V) |
Assay | (metals basis) |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P261-P264b-P270-P271-P301+P310-P304+P340-P311-P330-P501c
H301+H331
DOTInformation : Hazard Class: 6.1
TSCA : No
Recommended Storage : Ambient temperatures