Alfa Aesar™ Rubidium hydroxide, 50% w/w aq. soln., 99.6+% (metals basis)

Manufacturer: Alfa Aesar™ 10565NC

Quantity 10g

 View more versions of this product

Catalog No. AA10565NC

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 1310-82-3
    Linear Formula RbOH
    Formula Weight 102.48 (Anhydrous)
    MDL Number MFCD00011194
    UN Number UN2677
    Merck Index 14,8285
    Chemical Name or Material Rubidium hydroxide

    Specifications

    Formula Weight 102.48 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 1310-82-3
    UN Number UN2677
    Quantity 10g
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: RUBIDIUM HYDROXIDE SOLUTION
    Physical Form Liquid
    Merck Index 14,8285
    Concentration 50% w/w aq. soln.
    RTECS Number VL8750000
    TSCA TSCA
    Linear Formula RbOH
    MDL Number MFCD00011194
    Color Pale yellow
    Health Hazard 2 GHS H Statement
    H314-H302
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Chemical Name or Material Rubidium hydroxide
    Assay ≥99.6%, (metals basis)

    GHS H Statement
    H314-H302
    Causes severe skin burns and eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit