Alfa Aesar™ Rubidium hydroxide hydrate, 99% (metals basis)

Oxidative chlorination catalyst

Manufacturer: Alfa Aesar™ 1348106

Quantity 5g

 View more versions of this product

Catalog No. AA1348106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12026-05-0
    Linear Formula RbOH·xH2O
    Formula Weight 102.48 (Anhydrous)
    MDL Number MFCD00011194
    UN Number UN2678
    Merck Index 14,8285
    Chemical Name or Material Rubidium hydroxide hydrate

    Specifications

    Formula Weight 102.48 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Solubility Information Soluble in water, alcohol
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: RUBIDIUM HYDROXIDE
    MDL Number MFCD00011194
    Health Hazard 1 Danger
    CAS 12026-05-0
    Quantity 5g
    TSCA TSCA
    Physical Form Crystalline
    Linear Formula RbOH·xH2O
    UN Number UN2678
    Merck Index 14,8285
    Sensitivity Hygroscopic
    Color White to gray
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Chemical Name or Material Rubidium hydroxide hydrate
    Assay 99%, (metals basis)

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit