Alfa Aesar™ Rubidium iodate, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 3951709

Quantity 10g

 View more versions of this product

Catalog No. AA3951709

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 13446-76-9
    Formula Weight 260.37
    MDL Number MFCD00049524
    UN Number UN1479
    Synonym Rubidium iodine oxide
    Chemical Name or Material Rubidium iodate

    Specifications

    Formula Weight 260.37
    Health Hazard 3 GHS P Statement
    P221-P210-P284a-P220-P342+P311-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.Keep/Store away from clothing/combustible materials.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H272-H334-H317
    May intensify fire
    oxidizer.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.
    MDL Number MFCD00049524
    Synonym Rubidium iodine oxide
    Quantity 10g
    Mesh Size −4 Mesh
    CAS 13446-76-9
    UN Number UN1479
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    TSCA TSCA
    Health Hazard 1 Danger
    Chemical Name or Material Rubidium iodate
    Assay 99.9%, (metals basis)

    GHS H Statement
    H272-H334-H317
    May intensify fire
    oxidizer.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P221-P210-P284a-P220-P342+P311-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.Keep/Store away from clothing/combustible materials.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit