Rubidium Oxide, 99.9%, MP Biomedicals

$1570.35

Chemical Identifiers

Chemical Name or Material Rubidium Oxide
CAS [18088-11-4]
Formula Weight 186.94

  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  

ICN217876

MP Biomedicals
0521787601
1g
Each for $1,570.35
Check Availability
Add to Cart
Descriptions & Specifications
Safety and Handling

Related Products