Alfa Aesar™ Rubidium sulfate, Puratronic™, 99.975% (metals basis)

Manufacturer: Alfa Aesar™ 1287204

Quantity 2g

 View more versions of this product

Catalog No. AA1287204

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7488-54-2
    Formula Weight 267
    MDL Number MFCD00011190
    Chemical Name or Material Rubidium sulfate
    Grade Puratronic™

    Specifications

    Formula Weight 267
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    MDL Number MFCD00011190
    Melting Point 1050°C
    Quantity 2g
    Health Hazard 1 Warning
    Density 3.613g/mL
    TSCA TSCA
    Physical Form Crystalline
    Refractive Index 1.513
    CAS 7488-54-2
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Chemical Name or Material Rubidium sulfate
    Assay 99.975%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit