Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RUFY2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558931
Description
Immunogen sequence: EKAQMEAEDE DEKYLQECLS KSDSLQKQIS QKEKQLVQLE TDLKIEKEWR QTLQE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 95%.
RUFY2 (RUN And FYVE Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is RUNDC3B.
Specifications
RUFY2 | |
Polyclonal | |
Unconjugated | |
RUFY2 | |
2610111M19Rik; AI844453; antigen MU-RMS-40.17; Denn; Kiaa1537; Leucine zipper FYVE-finger protein; LZ-FYVE; Rab4-interacting protein related; RABIP4R; RUFY2; RUN and FYVE domain containing 2; RUN and FYVE domain-containing 2; RUN and FYVE domain-containing protein 2; Run- and FYVE-domain containing protein; ZFYVE13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55680 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WXA3 | |
RUFY2 | |
Recombinant protein corresponding to Human RUFY2. Recombinant protein control fragment (Product #RP-97227). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction